2-Hydroxy-5-nitropyridine 98%
Supplier: Apollo Scientific
Warning
Synonyms: 5-Nitro-2-pyridinol
OR4275-500G OR4275-1KG OR4275-50G
APOSOR4275-500GEA 174 EUR
APOSOR4275-500G APOSOR4275-1KG APOSOR4275-50G
2-Hydroxy-5-nitropyridine 98%
2-Hydroxy-5-nitropyridine
Formula: C₅H₄N₂O₃
MW: 140.1 g/mol
Melting Pt: 188…191 °C
Storage Temperature: Ambient
MDL Number: MFCD00006276
CAS Number: 5418-51-9
EINECS: 226-525-7