2-Hydroxyhippuric acid
Supplier: Apollo Scientific
Warning
Synonyms: N-(2-Hydroxybenzoyl)glycine
OR4276-1G OR4276-250MG
APOSOR4276-1GEA 49.3 EUR
APOSOR4276-1G APOSOR4276-250MG
2-Hydroxyhippuric acid
2-Hydroxyhippuric acid
Formula: C₉H₉NO₄
Melting Pt: 159…161 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002695
CAS Number: 487-54-7
EINECS: 207-661-6