cis-Aconitic acid 90%
Supplier: Apollo Scientific
Warning
Synonyms: (Z)-Prop-1-ene-1,2,3-tricarboxylic acid, (1Z)-Prop-1-ene-1,2,3-tricarboxylic acid, (Z)-Aconitic acid
OR4293-1G OR4293-250MG
APOSOR4293-1GEA 130 EUR
APOSOR4293-1G APOSOR4293-250MG
cis-Aconitic acid 90%
cis-Aconitic acid
Formula: C₆H₆O₆
MW: 174.11 g/mol
Melting Pt: 115…122 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00063184
CAS Number: 585-84-2
EINECS: 209-561-1