4'-Hydroxy-3'-nitroacetophenone 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1-(4-Hydroxy-3-nitrophenyl)-1-ethanone
OR4297-100G OR4297-25G
APOSOR4297-100GEA 0 EUR
APOSOR4297-100G APOSOR4297-25G
4'-Hydroxy-3'-nitroacetophenone 98%
4'-Hydroxy-3'-nitroacetophenone
Formula: C₈H₇NO₄
MW: 181.15
Melting Pt: 132…133 °C
Storage Temperature: Ambient
MDL Number: MFCD00017002
CAS Number: 6322-56-1