trans,trans-1,4-Diphenyl-1,3-butadiene 98%
Supplier: Apollo Scientific
Warning
Synonyms: (1E,3E)-1,4-Diphenylbuta-1,3-diene
OR4298-25G OR4298-5G
APOSOR4298-25GEA 40.6 EUR
APOSOR4298-25G APOSOR4298-5G
trans,trans-1,4-Diphenyl-1,3-butadiene 98%
trans,trans-1,4-Diphenyl-1,3-butadiene
Formula: C₁₆H₁₄
MW: 206.29 g/mol
Boiling Pt: 350 °C
Melting Pt: 150…152 °C
Storage Temperature: Ambient
MDL Number: MFCD00004791
CAS Number: 538-81-8
EINECS: 212-952-6