2,6-Dichloro-p-phenylenediamine 98%
Supplier: Apollo Scientific
Danger
Synonyms: 1,4-Diamino-2,6-dichlorobenzene, 2,6-Dichlorobenzene-1,4-diamine, 2,6-Dichloro-1,4-phenylenediamine
OR4321-1G OR4321-5G OR4321-10G
APOSOR4321-1GEA 21.9 EUR
APOSOR4321-1G APOSOR4321-5G APOSOR4321-10G
2,6-Dichloro-p-phenylenediamine 98%
2,6-Dichloro-p-phenylenediamine
Formula: C₆H₆Cl₂N₂
MW: 177.03 g/mol
Boiling Pt: 305
Melting Pt: 122…127 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00025369
CAS Number: 609-20-1
EINECS: 210-184-6