2-Bromobenzaldehyde ethylene acetal 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2-(2-Bromophenyl)-1,3-dioxolane
OR4329-100G OR4329-25G OR4329-500G
APOSOR4329-100GEA 132 EUR
APOSOR4329-100G APOSOR4329-25G APOSOR4329-500G
2-Bromobenzaldehyde ethylene acetal 98%
2-Bromobenzaldehyde ethylene acetal
Formula: C₉H₉BrO₂
MW: 229.07 g/mol
Boiling Pt: 126…127 °C/5mmHg
Density: 1.535 g/cm³ (25 °C)
Flash Pt: >121 °C
Storage Temperature: Ambient
MDL Number: MFCD00155124
CAS Number: 34824-58-3