2'-Bromoacetanilide
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-Bromoacetanilide, N-(2-Bromophenyl)acetamide
OR4339-100G
OR4339-25G
OR4339-5G
APOSOR4339-100GEA
79.9
EUR
APOSOR4339-100G
APOSOR4339-25G
APOSOR4339-5G
2'-Bromoacetanilide
2'-Bromoacetanilide
Formula:
H₃CC(O)NH(C₆H₄Br) MW: 214.06 g/mol Melting Pt: 96.5…100.5 °C Storage Temperature: Ambient |
MDL Number:
MFCD00099252 CAS Number: 614-76-6 |