4'-Chloroacetanilide
Supplier: Apollo Scientific
Warning
OR4345-100G OR4345-25G
APOSOR4345-100GEA 55.1 EUR
APOSOR4345-100G APOSOR4345-25G
4'-Chloroacetanilide
4'-Chloroacetanilide
Formula: C₈H₈ClNO
Boiling Pt: 333 °C
Melting Pt: 176…180 °C
Density: 1.385 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000612
CAS Number: 539-03-7
EINECS: 208-707-8