1-Chloro-3,5-dimethoxybenzene 98%
Supplier: Apollo Scientific
Warning
Synonyms: 5-Chloro-1,3-dimethoxybenzene
OR4352-25G OR4352-100G
APOSOR4352-25GEA 0 EUR
APOSOR4352-25G APOSOR4352-100G
1-Chloro-3,5-dimethoxybenzene 98%
1-Chloro-3,5-dimethoxybenzene
Formula: C₈H₉ClO₂
MW: 172.61 g/mol
Boiling Pt: 234 °C
Melting Pt: 32…36 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00008382
CAS Number: 7051-16-3
EINECS: 230-330-2