3-Chloro-4-hydroxybenzaldehyde 98%
Supplier: Apollo Scientific
Danger
OR4357-100G OR4357-10G OR4357-25G
APOSOR4357-100GEA 108 EUR
APOSOR4357-100G APOSOR4357-10G APOSOR4357-25G
3-Chloro-4-hydroxybenzaldehyde 98%
3-Chloro-4-hydroxybenzaldehyde
Formula: C₇H₅ClO₂
MW: 156.57 g/mol
Boiling Pt: 150 °C/15mmHg
Melting Pt: 133…137 °C
Density: 1.0404
Storage Temperature: Ambient
MDL Number: MFCD00016981
CAS Number: 2420-16-8