Methyl-2-chloro-6-methoxynicotinate 97%
Supplier: Apollo Scientific
Warning
Synonyms: Methyl 2-chloro-6-methoxynicatinate, Methyl 2-chloro-6-methoxynicotinate
OR43596-5G
APOSOR43596-5GEA 143 EUR
APOSOR43596-5G
Methyl-2-chloro-6-methoxynicotinate 97%
Methyl 2-chloro-6-methoxynicotinate
Formula: C₈H₈ClNO₃
MW: 201.61 g/mol
Melting Pt: 67…69 °C
Storage Temperature: Ambient
MDL Number: MFCD08275100
CAS Number: 95652-77-0