3-Nitrobenzoic acid 99%
Supplier: Apollo Scientific
Warning
Synonyms: m-Nitrobenzoic acid
OR4367-1KG OR4367-500G OR4367-5KG
APOSOR4367-1KGEA 33.5 EUR
APOSOR4367-1KG APOSOR4367-500G APOSOR4367-5KG
3-Nitrobenzoic acid 99%
3-Nitrobenzoic acid
Formula: C₇H₅NO₄
MW: 167.12 g/mol
Boiling Pt: 341
Melting Pt: 139…143 °C
Density: 1.494 g/cm³ (25 °C)
Flash Pt: 190 °C
Storage Temperature: Ambient
MDL Number: MFCD00007251
CAS Number: 121-92-6
EINECS: 204-508-5
Merck Index: 12,06686