3,4,5-Trimethoxybenzhydrazide 98%
Supplier: Apollo Scientific
Warning
OR4394-25G OR4394-5G
APOSOR4394-25GEA 63 EUR
APOSOR4394-25G APOSOR4394-5G
3,4,5-Trimethoxybenzhydrazide 98%
3,4,5-Trimethoxybenzhydrazide
Formula: C₁₀H₁₄N₂O₄
Melting Pt: 169…170 °C
Storage Temperature: Ambient
MDL Number: MFCD00014758
CAS Number: 3291-03-0
EINECS: 221-954-6