4-(2-Methylthiazol-4-yl)benzenesulphonyl chloride 97%
Supplier: Apollo Scientific
Danger
OR4395-1G OR4395-250MG
APOSOR4395-1GEA 174 EUR
APOSOR4395-1G APOSOR4395-250MG
4-(2-Methylthiazol-4-yl)benzenesulphonyl chloride 97%
4-(2-Methylthiazol-4-yl)benzenesulphonyl chloride
Formula: C₁₀H₈ClNO₂S₂
MW: 273.76 g/mol
Melting Pt: 108…112 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07368551
CAS Number: 852180-73-5
UN: 3261
ADR: 8 III