3-Amino-2-chloro-6-methylphenol
Supplier: Apollo Scientific
Warning
Synonyms: 2-Chloro-4-methyl-3-hydroxyaniline, 5-Amino-6-chloro-o-cresol
OR4402-25G OR4402-5G
APOSOR4402-25GEA 33.5 EUR
APOSOR4402-25G APOSOR4402-5G
3-Amino-2-chloro-6-methylphenol
3-Amino-2-chloro-6-methylphenol
Formula: C₇H₈ClNO
MW: 157.60 g/mol
Melting Pt: 75…77 °C
Storage Temperature: Ambient
MDL Number: MFCD03094650
CAS Number: 84540-50-1
EINECS: 283-144-9