2,5-Dibromo-3-formylpyridine
Supplier: Apollo Scientific
Synonyms: 2,5-Dibromonicotinaldehyde
OR4434-1G OR4434-250MG OR4434-500MG OR4434-2.5G
APOSOR4434-1GEA 314 EUR
APOSOR4434-1G APOSOR4434-250MG APOSOR4434-500MG APOSOR4434-2.5G
2,5-Dibromo-3-formylpyridine
2,5-Dibromo-3-formylpyridine
Formula: C₆H₃Br₂NO
MW: 264.9021 g/mol
Melting Pt: 75.5…77 °C
Storage Temperature: Ambient
MDL Number: MFCD07368560
CAS Number: 852181-11-4