2-Chloroaniline 98+%
Supplier: Apollo Scientific
Danger
OR4444-500G OR4444-100G OR4444-1KG
APOSOR4444-500GEA 109 EUR
APOSOR4444-500G APOSOR4444-100G APOSOR4444-1KG
2-Chloroaniline 98+%
2-Chloroaniline
Formula: C₆H₆ClN
MW: 127.57 g/mol
Boiling Pt: 209 °C
Melting Pt: –3 °C
Density: 1.22 g/cm³ (25 °C)
Flash Pt: 104 °C
Storage Temperature: Ambient
MDL Number: MFCD00007656
CAS Number: 95-51-2
UN: 2019
ADR: 6.1,II