4-Chlorobenzenesulphonyl chloride 99%
Supplier: Apollo Scientific
Danger
Synonyms: 4-Chlorobenzene-1-sulphonyl chloride
OR4476-100G OR4476-500G OR4476-25G
APOSOR4476-100GEA 37.7 EUR
APOSOR4476-100G APOSOR4476-500G APOSOR4476-25G
4-Chlorobenzenesulphonyl chloride 99%
4-Chlorobenzenesulphonyl chloride
Formula: C₆H₄Cl₂O₂S
MW: 211.07 g/mol
Boiling Pt: 141 °C/15mmHg
Melting Pt: 50…52 °C
Flash Pt: 108 °C
Storage Temperature: Ambient
MDL Number: MFCD00007439
CAS Number: 98-60-2
EINECS: 202-685-3
UN: 3261
ADR: 8 II