4-Bromo-2,6-dichlorobenzenesulphonyl chloride 96%
Supplier: Apollo Scientific
Danger
Synonyms: 4-Bromo-2,6-dichlorobenzene-1-sulphonyl chloride
OR4495-5G OR4495-1G
APOSOR4495-5GEA 58 EUR
APOSOR4495-5G APOSOR4495-1G
4-Bromo-2,6-dichlorobenzenesulphonyl chloride 96%
4-Bromo-2,6-dichlorobenzenesulphonyl chloride
Formula: C₆H₂BrCl₃O₂S
MW: 324.41 g/mol
Melting Pt: 46…50 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD03094651
CAS Number: 351003-54-8
UN: 3261
ADR: 8,II