6-Bromo-2-(chloromethyl)-3-nitroimidazo[1,2-a]pyridine
Supplier: Apollo Scientific
OR450074-1G
OR450074-250MG
OR450074-100MG
OR450074-500MG
OR450074-2.5G
OR450074-5G
APOSOR450074-1GEA
589
EUR
APOSOR450074-1G
APOSOR450074-250MG
APOSOR450074-100MG
APOSOR450074-500MG
APOSOR450074-2.5G
APOSOR450074-5G
6-Bromo-2-(chloromethyl)-3-nitroimidazo[1,2-a]pyridine
6-Bromo-2-(chloromethyl)-3-nitroimidazo[1,2-a]pyridine
Formula:
C₈H₅BrClN₃O₂ MW: 290.51 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08704640 CAS Number: 708213-35-8 |