2-Methyl-3-nitroimidazo[1,2-a]pyridine
Supplier: Apollo Scientific
OR450076-1G OR450076-250MG OR450076-500MG
APOSOR450076-1GEA 171 EUR
APOSOR450076-1G APOSOR450076-250MG APOSOR450076-500MG
2-Methyl-3-nitroimidazo[1,2-a]pyridine
2-Methyl-3-nitroimidazo[1,2-a]pyridine
Formula: C₈H₇N₃O₂
Storage Temperature: Ambient
MDL Number: MFCD09800612
CAS Number: 34165-09-8