6-Chloro-m-anisidine
Supplier: Apollo Scientific
Danger
Synonyms: 6-Chloro-3-anisidine, 2-Chloro-5-methoxyaniline, 3-Amino-4-chloroanisole
OR4502-5G OR4502-1G
APOSOR4502-5GEA 29 EUR
APOSOR4502-5G APOSOR4502-1G
6-Chloro-m-anisidine
6-Chloro-m-anisidine
Formula: C₇H₈ClNO
MW: 157.6 g/mol
Boiling Pt: 135…137 °C/12mmHg
Melting Pt: 27 °C
Storage Temperature: Ambient
MDL Number: MFCD00047830
CAS Number: 2401-24-3
EINECS: 219-277-6
UN: 2233
ADR: 6.1,III