1-Chloro-3,5-dibromobenzene
Supplier: Apollo Scientific
Warning
Synonyms: 3,5-Dibromochlorobenzene, 1,3-Dibromo-5-chlorobenzene
OR4505-100G OR4505-25G OR4505-5G
APOSOR4505-100GEA 74.1 EUR
APOSOR4505-100G APOSOR4505-25G APOSOR4505-5G
1-Chloro-3,5-dibromobenzene
1-Chloro-3,5-dibromobenzene
Formula: C₆H₃Br₂Cl
MW: 270.35 g/mol
Boiling Pt: 256 °C
Melting Pt: 91…98 °C
Storage Temperature: Ambient
MDL Number: MFCD00070765
CAS Number: 14862-52-3