6-Amino-3-bromoquinoline
Supplier: Apollo Scientific
Warning
Synonyms: 3-Bromoquinolin-6-amine
OR45086-1G OR45086-250MG
APOSOR45086-1GEA 185 EUR
APOSOR45086-1G APOSOR45086-250MG
6-Amino-3-bromoquinoline
6-Amino-3-bromoquinoline
Formula: C₉H₇BrN₂
MW: 223.07 g/mol
Melting Pt: 154…155 °C
Storage Temperature: Ambient
MDL Number: MFCD07757603
CAS Number: 7101-96-4