4-Tetrahydrothiopyranone-1,1-dioxide
Supplier: Apollo Scientific
OR451043-5G OR451043-1G OR451043-25G
APOSOR451043-5GEA 101 EUR
APOSOR451043-5G APOSOR451043-1G APOSOR451043-25G
4-Tetrahydrothiopyranone-1,1-dioxide
4-Tetrahydrothiopyranone-1,1-dioxide
Formula: C₅H₈O₃S
Storage Temperature: Ambient
MDL Number: MFCD00460698
CAS Number: 17396-35-9