Quinoline 98%
Supplier: Apollo Scientific
Danger
OR45105-100G OR45105-500G OR45105-250G OR45105-25G
APOSOR45105-100GEA 69.6 EUR
APOSOR45105-100G APOSOR45105-500G APOSOR45105-250G APOSOR45105-25G
Quinoline 98%
Quinoline
Formula: C₉H₇N
MW: 129.16 g/mol
Boiling Pt: 236…238 °C
Melting Pt: –17 to –13 °C
Density: 1.093 g/cm³ (25 °C)
Flash Pt: 101 °C
Storage Temperature: Ambient
MDL Number: MFCD00006736
CAS Number: 91-22-5
EINECS: 202-051-6
UN: 2656
ADR: 6.1,III