3-Bromo-2,4,6-trimethylaniline 98%
Supplier: Apollo Scientific
Warning
OR451307-25G OR451307-5G
APOSOR451307-25GEA 116 EUR
APOSOR451307-25G APOSOR451307-5G
3-Bromo-2,4,6-trimethylaniline 98%
3-Bromo-2,4,6-trimethylaniline
Formula: C₉H₁₂BrN
MW: 214.11 g/mol
Boiling Pt: 172…173 °C
Melting Pt: 34.5…39.3 °C
Density: 1.313 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00015476
CAS Number: 82842-52-2