2,6-Dimethylthiophenol 95+%
Supplier: Apollo Scientific
Warning
Synonyms: 1,3-Dimethyl-2-mercaptobenzene, 2-Mercapto-m-xylene, 2,6-Dimethylbenzene-1-thiol, 2,6-Dimethylbenzenethiol
OR451316-100G OR451316-25G OR451316-5G
APOSOR451316-100GEA 365 EUR
APOSOR451316-100G APOSOR451316-25G APOSOR451316-5G
2,6-Dimethylthiophenol 95+%
2,6-Dimethylthiophenol
Formula: C₈H₁₀S
MW: 138.23 g/mol
Boiling Pt: 122 °C
Density: 1.038 g/cm³ (25 °C)
Flash Pt: 86 °C
Storage Temperature: Ambient
MDL Number: MFCD00010021
CAS Number: 118-72-9
UN: 3334
ADR: 9 III