Furfuryl methyl sulphide 97+%
Supplier: Apollo Scientific
Warning
Synonyms: 2-((Methylthio)methyl)furan
OR451317-100G OR451317-25G
APOSOR451317-100GEA 62.5 EUR
APOSOR451317-100G APOSOR451317-25G
Furfuryl methyl sulphide 97+%
Furfuryl methyl sulphide
Formula: C₆H₈OS
MW: 128.19 g/mol
Boiling Pt: 64…65 °C
Density: 1.07 g/cm³ (25 °C)
Flash Pt: 63 °C
Storage Temperature: Ambient
MDL Number: MFCD00009604
CAS Number: 1438-91-1
UN: 3334
ADR: 9,III