1-Chloro-6-iodohexane 95%
Supplier: Apollo Scientific
Warning
OR451323-25G OR451323-5G OR451323-100G
APOSOR451323-25GEA 56.7 EUR
APOSOR451323-25G APOSOR451323-5G APOSOR451323-100G
1-Chloro-6-iodohexane 95%
1-Chloro-6-iodohexane
Formula: C₆H₁₂ClI
MW: 246.52 g/mol
Boiling Pt: 110 °C
Density: 1.623 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00039417
CAS Number: 34683-73-3