o-Toluamide 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Methylbenzamide
OR451329-25G OR451329-100G
APOSOR451329-25GEA 23.2 EUR
APOSOR451329-25G APOSOR451329-100G
o-Toluamide 98%
o-Toluamide
Formula: C₈H₉NO
MW: 135.17 g/mol
Melting Pt: 141…142 °C
Storage Temperature: Ambient
MDL Number: MFCD00007982
CAS Number: 527-85-5