1-(4-Bromo-1-methyl-1H-pyrazol-5-yl)ethanone
Supplier: Apollo Scientific
Warning
OR451418-1G OR451418-5G
APOSOR451418-1GEA 34.8 EUR
APOSOR451418-1G APOSOR451418-5G
1-(4-Bromo-1-methyl-1H-pyrazol-5-yl)ethanone
1-(4-Bromo-1-methyl-1H-pyrazol-5-yl)ethanone
Formula: C₆H₇BrN₂O
MW: 203.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04968842
CAS Number: 925200-47-1