3-Methyl-1H-pyrazol-4-amine hydrochloride
Supplier: Apollo Scientific
Warning
Synonyms: 3-Methyl-1H-pyrazol-4-ylamine hydrochloride
OR451430-1G OR451430-25G OR451430-5G OR451430-10G
APOSOR451430-1GEA 203 EUR
APOSOR451430-1G APOSOR451430-25G APOSOR451430-5G APOSOR451430-10G
3-Methyl-1H-pyrazol-4-amine hydrochloride
3-Methyl-1H-pyrazol-4-amine hydrochloride
Formula: C₄H₈ClN₃
MW: 133.58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16613948
CAS Number: 113140-10-6