3-Methylisoxazole-5-carbonitrile
Supplier: Apollo Scientific
Warning
OR451449-1G OR451449-5G
APOSOR451449-1GEA 65.4 EUR
APOSOR451449-1G APOSOR451449-5G
3-Methylisoxazole-5-carbonitrile
3-Methylisoxazole-5-carbonitrile
Formula: C₅H₄N₂O
Storage Temperature: Ambient
MDL Number: MFCD14702909
CAS Number: 65735-07-1