Ethyl 1,2,4-triazine-3-carboxylate, Technical Grade
Supplier: Apollo Scientific
Warning
Synonyms: Ethyl 1,2,4-triazine-3-carboxylate
OR45145-1G OR45145-250MG OR45145-50MG OR45145-100MG OR45145-500MG
APOSOR45145-1GEA 1040 EUR
APOSOR45145-1G APOSOR45145-250MG APOSOR45145-50MG APOSOR45145-100MG APOSOR45145-500MG
Ethyl 1,2,4-triazine-3-carboxylate, Technical Grade
Ethyl-1,2,4-triazine-3-carboxylate
Formula: C₆H₇N₃O₂
MW: 153.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10566533
CAS Number: 6498-02-8