4-Aminopyrazolo[5,1-c][1,2,4]triazine-3-carbonitrile
Supplier: Apollo Scientific
|
Warning
|
OR451458-1G
OR451458-5G
APOSOR451458-1GEA
172
EUR
APOSOR451458-1G
APOSOR451458-5G
4-Aminopyrazolo[5,1-c][1,2,4]triazine-3-carbonitrile
4-Aminopyrazolo[5,1-c][1,2,4]triazine-3-carbonitrile
Formula:
C₆H₄N₆ MW: 160.14 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03829962 CAS Number: 58390-42-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...