2-Bromo-4,5-dimethoxybenzoic acid
Supplier: Apollo Scientific
Warning
Synonyms: 6-Bromoveratric acid
OR4517-1G OR4517-25G OR4517-5G
APOSOR4517-1GEA 21.9 EUR
APOSOR4517-1G APOSOR4517-25G APOSOR4517-5G
2-Bromo-4,5-dimethoxybenzoic acid
2-Bromo-4,5-dimethoxybenzoic acid
Formula: C₉H₉BrO₄
MW: 261.07 g/mol
Melting Pt: 188…190 °C
Storage Temperature: Ambient
MDL Number: MFCD00017542
CAS Number: 6286-46-0