2-Bromo-5-chlorothiophene 98%
Supplier: Apollo Scientific
Warning
OR4520-100G OR4520-25G OR4520-5G
APOSOR4520-100GEA 120 EUR
APOSOR4520-100G APOSOR4520-25G APOSOR4520-5G
2-Bromo-5-chlorothiophene 98%
2-Bromo-5-chlorothiophene
Formula: C₄H₂BrClS
MW: 197.48 g/mol
Boiling Pt: 69…71 °C/18mmHg
Density: 1.803 g/cm³ (25 °C)
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00014523
CAS Number: 2873-18-9
EINECS: 220-708-5