Boc-5-bromo-DL-tryptophan 97%
Supplier: Apollo Scientific
OR452005-1G OR452005-250MG OR452005-500MG
APOSOR452005-1GEA 491 EUR
APOSOR452005-1G APOSOR452005-250MG APOSOR452005-500MG
Boc-5-bromo-DL-tryptophan 97%
Boc-5-bromo-DL-tryptophan
Formula: C₁₆H₁₉BrN₂O₄
MW: 383.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02682356
CAS Number: 67308-26-3