3-Dimethylaminomethylbenzoic acid hydrochloride 97%
Supplier: Apollo Scientific
OR452054-1G OR452054-5G
APOSOR452054-1GEA 456 EUR
APOSOR452054-1G APOSOR452054-5G
3-Dimethylaminomethylbenzoic acid hydrochloride 97%
3-Dimethylaminomethylbenzoic acid hydrochloride
Formula: C₁₀H₁₄ClNO₂
MW: 215.68 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09997603
CAS Number: 155412-73-0