2,5-Dibromobenzoic acid
Supplier: Apollo Scientific
Warning
OR4523-25G OR4523-5G
APOSOR4523-25GEA 45.1 EUR
APOSOR4523-25G APOSOR4523-5G
2,5-Dibromobenzoic acid
2,5-Dibromobenzoic acid
Formula: C₇H₄Br₂O₂
MW: 279.92 g/mol
Melting Pt: 156…159 °C
Storage Temperature: Ambient
MDL Number: MFCD00016494
CAS Number: 610-71-9
EINECS: 210-234-7