2,4-Dichloro-5-methoxyaniline
Supplier: Apollo Scientific
Warning
Synonyms: 5-Amino-2,4-dichloroanisole, 4,6-Dichloro-m-anisidine
OR4527-5G OR4527-100G OR4527-25G
APOSOR4527-5GEA 21.9 EUR
APOSOR4527-5G APOSOR4527-100G APOSOR4527-25G
2,4-Dichloro-5-methoxyaniline
2,4-Dichloro-5-methoxyaniline
Formula: C₇H₇Cl₂NO
Melting Pt: 50…51 °C
Storage Temperature: Ambient
MDL Number: MFCD00974410
CAS Number: 98446-49-2
UN: 2811
ADR: 6.1,III