3-Chlorobenzoyl chloride 99%
Supplier: Apollo Scientific
Danger
Synonyms: m-Chlorobenzoyl chloride, 3-Chlorobenzoylchloride
OR4528-100G OR4528-25G OR4528-500G
APOSOR4528-100GEA 43.5 EUR
APOSOR4528-100G APOSOR4528-25G APOSOR4528-500G
3-Chlorobenzoyl chloride 99%
3-Chlorobenzoyl chloride
Formula: C₇H₄Cl₂O
MW: 175.01 g/mol
Boiling Pt: 224…225 °C
Density: 1.368 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00000671
CAS Number: 618-46-2
EINECS: 210-552-6
UN: 3265
ADR: 8,II