1-Iodo-3-nitrobenzene 95%
Supplier: Apollo Scientific
Warning
OR4539-100G OR4539-1KG OR4539-500G
APOSOR4539-100GEA 286 EUR
APOSOR4539-100G APOSOR4539-1KG APOSOR4539-500G
1-Iodo-3-nitrobenzene 95%
1-Iodo-3-nitrobenzene
Formula: C₆H₄INO₂
MW: 249.01 g/mol
Boiling Pt: 280 °C
Melting Pt: 35…38 °C
Flash Pt: 71 °C
Storage Temperature: Ambient
MDL Number: MFCD00007218
CAS Number: 645-00-1
UN: 2926
ADR: 4.1+6.1 III