4-Methoxyindole 98%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Methoxy-1H-indole, Indol-4-yl methyl ether
OR4545-1G OR4545-25G OR4545-5G
APOSOR4545-1GEA 23.2 EUR
APOSOR4545-1G APOSOR4545-25G APOSOR4545-5G
4-Methoxyindole 98%
4-Methoxyindole
Formula: C₉H₉NO
MW: 147.18 g/mol
Boiling Pt: 181…183 °C/24mmHg
Melting Pt: 69…71 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009737
CAS Number: 4837-90-5