4-Allyl-3-thiosemicarbazide
Supplier: Apollo Scientific
Warning
OR4563-1G OR4563-5G
APOSOR4563-1GEA 58 EUR
APOSOR4563-1G APOSOR4563-5G
4-Allyl-3-thiosemicarbazide
4-Allyl-3-thiosemicarbazide
Formula: C₄H₉N₃S
Melting Pt: 93…96 °C
Storage Temperature: Ambient
MDL Number: MFCD00007619
CAS Number: 3766-55-0