(Methylthio)acetic acid 99%
Supplier: Apollo Scientific
Warning
Synonyms: (Methylthio)ethansäure, 2-(Methylthio)acetic acid
OR4573-100G OR4573-25G
APOSOR4573-100GEA 522 EUR
APOSOR4573-100G APOSOR4573-25G
(Methylthio)acetic acid 99%
(Methylthio)acetic acid
Formula: C₃H₆O₂S
MW: 106.15 g/mol
Boiling Pt: 130…137 °C/27mmHg
Melting Pt: 13…14 °C
Density: 1.219 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00075444
CAS Number: 2444-37-3