5-Bromo-2-iodobenzoic acid 97+%
Supplier: Apollo Scientific
Warning
OR4590-100G OR4590-25G OR4590-5G
APOSOR4590-100GEA 71.2 EUR
APOSOR4590-100G APOSOR4590-25G APOSOR4590-5G
5-Bromo-2-iodobenzoic acid 97+%
5-Bromo-2-iodobenzoic acid
Formula: C₇H₄BrIO₂
MW: 326.92 g/mol
Melting Pt: 159…163 °C
Storage Temperature: Ambient
MDL Number: MFCD00079717
CAS Number: 21740-00-1