2-Chlorobutane
Supplier: Apollo Scientific
Danger
OR460003-100ML OR460003-25ML
APOSOR460003-100MLEA 72.7 EUR
APOSOR460003-100ML APOSOR460003-25ML
2-Chlorobutane
2-Chlorobutane
Formula: C₄H₉Cl
Boiling Pt: 68…70 °C
Melting Pt: –140 °C
Density: 0.873 g/cm³ (25 °C)
Flash Pt: –15 °C
Storage Temperature: Ambient
MDL Number: MFCD00000871
CAS Number: 78-86-4
EINECS: 201-151-7
UN: 1127
ADR: 3,II